Anti-MYF6 Mouse Monoclonal Antibody [clone: 13B546]

Lieferant: United States Biological
USBI030468EA 437 EUR
USBI030468
Anti-MYF6 Mouse Monoclonal Antibody [clone: 13B546]
Antikörper
Type: Primary
Antigen: MYF6 (myogenic factor 6 (herculin))
Clonality: Monoclonal
Clone: 13B546
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR