Anti-NKX2-5 Mouse Monoclonal Antibody [clone: 13B572]

Lieferant: United States Biological
USBI030494EA 412 EUR
USBI030494
Anti-NKX2-5 Mouse Monoclonal Antibody [clone: 13B572]
Antikörper
Type: Primary
Antigen: NKX2-5 (NK2 homeobox 5)
Clonality: Monoclonal
Clone: 13B572
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR