Anti-CDKN2A Mouse Monoclonal Antibody [clone: 13B594]

Lieferant: United States Biological
USBI030516EA 462 EUR
USBI030516
Anti-CDKN2A Mouse Monoclonal Antibody [clone: 13B594]
Antikörper
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000

Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: Monoclonal
Clone: 13B594
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR