Anti-PAX5 Mouse Monoclonal Antibody [clone: 13B606]

Lieferant: United States Biological
USBI030528EA 437 EUR
USBI030528
Anti-PAX5 Mouse Monoclonal Antibody [clone: 13B606]
Antikörper
Type: Primary
Antigen: PAX5 (paired box 5)
Clonality: Monoclonal
Clone: 13B606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR