Anti-PKN2 Mouse Monoclonal Antibody [clone: 13B642]

Lieferant: United States Biological
USBI030564EA 590 EUR
USBI030564
Anti-PKN2 Mouse Monoclonal Antibody [clone: 13B642]
Antikörper
Type: Primary
Antigen: PKN2 (protein kinase N2)
Clonality: Monoclonal
Clone: 13B642
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR