Anti-PTK7 Mouse Monoclonal Antibody [clone: 13B657]

Lieferant: United States Biological
USBI030579EA 412 EUR
USBI030579
Anti-PTK7 Mouse Monoclonal Antibody [clone: 13B657]
Antikörper
Type: Primary
Antigen: PTK7 (protein tyrosine kinase 7)
Clonality: Monoclonal
Clone: 13B657
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR