Anti-PTK2B Mouse Monoclonal Antibody [clone: 13B659]

Lieferant: United States Biological
USBI030581EA 567 EUR
USBI030581
Anti-PTK2B Mouse Monoclonal Antibody [clone: 13B659]
Antikörper
Type: Primary
Antigen: PTK2B (protein tyrosine kinase 2 beta)
Clonality: Monoclonal
Clone: 13B659
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR