Anti-RAF1 Mouse Monoclonal Antibody [clone: 13B667]

Lieferant: United States Biological
USBI030589EA 590 EUR
USBI030589
Anti-RAF1 Mouse Monoclonal Antibody [clone: 13B667]
Antikörper
Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: Monoclonal
Clone: 13B667
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR