Anti-RET Mouse Monoclonal Antibody [clone: 13B676]

Lieferant: United States Biological
USBI030598EA 526 EUR
USBI030598
Anti-RET Mouse Monoclonal Antibody [clone: 13B676]
Antikörper
Type: Primary
Antigen: RET (ret proto-oncogene)
Clonality: Monoclonal
Clone: 13B676
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR