Anti-SRY Mouse Monoclonal Antibody [clone: 13B737]

Lieferant: United States Biological
USBI030659EA 437 EUR
USBI030659
Anti-SRY Mouse Monoclonal Antibody [clone: 13B737]
Antikörper
Type: Primary
Antigen: SRY (sex determining region Y)
Clonality: Monoclonal
Clone: 13B737
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR