Anti-TDGF1 Mouse Monoclonal Antibody [clone: 13B758]

Lieferant: United States Biological
USBI030680EA 414 EUR
USBI030680
Anti-TDGF1 Mouse Monoclonal Antibody [clone: 13B758]
Antikörper
Type: Primary
Antigen: TDGF1 (teratocarcinoma-derived growth factor 1)
Clonality: Monoclonal
Clone: 13B758
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR