Anti-THPO Mouse Monoclonal Antibody [clone: 13B764]

Lieferant: United States Biological
USBI030686EA 437 EUR
USBI030686
Anti-THPO Mouse Monoclonal Antibody [clone: 13B764]
Antikörper
Type: Primary
Antigen: THPO (thrombopoietin)
Clonality: Monoclonal
Clone: 13B764
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR