Anti-UBA7 Mouse Monoclonal Antibody [clone: 13B790]

Lieferant: United States Biological
USBI030712EA 412 EUR
USBI030712
Anti-UBA7 Mouse Monoclonal Antibody [clone: 13B790]
Antikörper
Type: Primary
Antigen: UBA7 (ubiquitin-like modifier activating enzyme 7)
Clonality: Monoclonal
Clone: 13B790
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR