Anti-ACOX1 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 153CT43.1.1]

Lieferant: United States Biological
USBI030738-FITCEA 619 EUR
USBI030738-FITC
Anti-ACOX1 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 153CT43.1.1]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ACOX1 (acyl-CoA oxidase 1, palmitoyl)
Clonality: Monoclonal
Clone: 153CT43.1.1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR