Anti-AGT Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 656CT5.8.4]

Lieferant: United States Biological
USBI030750-FITCEA 619 EUR
USBI030750-FITC
Anti-AGT Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 656CT5.8.4]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: Monoclonal
Clone: 656CT5.8.4
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR