Anti-ALB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 500CT2.1.4]

Lieferant: United States Biological
USBI030760-HRPEA 619 EUR
USBI030760-HRP
Anti-ALB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 500CT2.1.4]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ALB (albumin)
Clonality: Monoclonal
Clone: 500CT2.1.4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR