Anti-ALDH1A1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 152CT1.2.2]
Lieferant: United States Biological
USBI030762-APEA 852 EUR
USBI030762-AP
Anti-ALDH1A1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 152CT1.2.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: Monoclonal
Clone: 152CT1.2.2
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human