Anti-ALDH1A1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 152CT1.2.2]

Lieferant: United States Biological
USBI030762-HRPEA 619 EUR
USBI030762-HRP
Anti-ALDH1A1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 152CT1.2.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: Monoclonal
Clone: 152CT1.2.2
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR