Anti-ALDH2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 138CT22.3.8]

Lieferant: United States Biological
USBI030763-FITCEA 619 EUR
USBI030763-FITC
Anti-ALDH2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 138CT22.3.8]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality: Monoclonal
Clone: 138CT22.3.8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR