Anti-APOA1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 464CT10.4.4]

Lieferant: United States Biological
USBI030784-HRPEA 619 EUR
USBI030784-HRP
Anti-APOA1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 464CT10.4.4]
Antikörper
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: Monoclonal
Clone: 464CT10.4.4
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR