Anti-BAX Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 557CT5.2.1]

Lieferant: United States Biological
USBI030802-HRPEA 619 EUR
USBI030802-HRP
Anti-BAX Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 557CT5.2.1]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: Monoclonal
Clone: 557CT5.2.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR