Anti-BMI1 Mouse Monoclonal Antibody [clone: 282CT3.7.6]

Lieferant: United States Biological
USBI030810EA 587 EUR
USBI030810
Anti-BMI1 Mouse Monoclonal Antibody [clone: 282CT3.7.6]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250; Immunofluorescence: 1:10-50

Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: Monoclonal
Clone: 282CT3.7.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR