Anti-REL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 318CT41.3.2]

Lieferant: United States Biological
USBI030818-APCEA 619 EUR
USBI030818-APC
Anti-REL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 318CT41.3.2]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:120-1000

Type: Primary
Antigen: REL (v-rel avian reticuloendotheliosis viral oncogene homolog)
Clonality: Monoclonal
Clone: 318CT41.3.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR