Anti-CAT Mouse Monoclonal Antibody [clone: 626CT4.3.8]

Lieferant: United States Biological
USBI030832EA 495 EUR
USBI030832
Anti-CAT Mouse Monoclonal Antibody [clone: 626CT4.3.8]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: CAT (catalase)
Clonality: Monoclonal
Clone: 626CT4.3.8
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR