Anti-VCAM1 Mouse Monoclonal Antibody [clone: 582CT25.6.4]

Lieferant: United States Biological
USBI030836EA 495 EUR
USBI030836
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 582CT25.6.4]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 582CT25.6.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR