Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 7G7B6]

Lieferant: United States Biological
USBI030844-FITCEA 562 EUR
USBI030844-FITC
Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 7G7B6]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250

Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: Monoclonal
Clone: 7G7B6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR