Anti-CD40 Mouse Monoclonal Antibody [clone: 647CT13.2.4]

Lieferant: United States Biological
USBI030851EA 495 EUR
USBI030851
Anti-CD40 Mouse Monoclonal Antibody [clone: 647CT13.2.4]
Antikörper
Recommended dilution: Western Blot: 1:100-1:1600

Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: Monoclonal
Clone: 647CT13.2.4
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR