Anti-CDC25C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 233CT9.6.6]

Lieferant: United States Biological
USBI030866-APCEA 619 EUR
USBI030866-APC
Anti-CDC25C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 233CT9.6.6]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: Monoclonal
Clone: 233CT9.6.6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR