Anti-CDCA8 Mouse Monoclonal Antibody [clone: 181CT41.2.3]

Lieferant: United States Biological
USBI030867EA 495 EUR
USBI030867
Anti-CDCA8 Mouse Monoclonal Antibody [clone: 181CT41.2.3]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:2000-16000

Type: Primary
Antigen: CDCA8 (cell division cycle associated 8)
Clonality: Monoclonal
Clone: 181CT41.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR