Anti-CDKN1A Mouse Monoclonal Antibody [clone: 592CT11.4.4]

Lieferant: United States Biological
USBI030871EA 587 EUR
USBI030871
Anti-CDKN1A Mouse Monoclonal Antibody [clone: 592CT11.4.4]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: CDKN1A (cyclin-dependent kinase inhibitor 1A (p21, Cip1))
Clonality: Monoclonal
Clone: 592CT11.4.4
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR