Anti-CHEK1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 2G1D5]

Lieferant: United States Biological
USBI030874-HRPEA 619 EUR
USBI030874-HRP
Anti-CHEK1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 2G1D5]
Antikörper
Recommended dilution: ELISA: 1:10000; Western Blot: 1:1000-4000

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: Monoclonal
Clone: 2G1D5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR