Anti-CNOT4 Mouse Monoclonal Antibody [clone: 343CT14.6.5]

Lieferant: United States Biological
USBI030880EA 495 EUR
USBI030880
Anti-CNOT4 Mouse Monoclonal Antibody [clone: 343CT14.6.5]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-8000

Type: Primary
Antigen: CNOT4 (CCR4-NOT transcription complex, subunit 4)
Clonality: Monoclonal
Clone: 343CT14.6.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR