Anti-CSNK2B Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 260CT10.1.1]

Lieferant: United States Biological
USBI030892-APEA 619 EUR
USBI030892-AP
Anti-CSNK2B Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 260CT10.1.1]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: CSNK2B (casein kinase 2, beta polypeptide)
Clonality: Monoclonal
Clone: 260CT10.1.1
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR