Anti-CYP3A4 Mouse Monoclonal Antibody [clone: 501CT16.1.1]

Lieferant: United States Biological
USBI030906EA 420 EUR
USBI030906
Anti-CYP3A4 Mouse Monoclonal Antibody [clone: 501CT16.1.1]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: CYP3A4 (cytochrome P450, family 3, subfamily A, polypeptide 4)
Clonality: Monoclonal
Clone: 501CT16.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR