Anti-DPF2 Mouse Monoclonal Antibody [clone: 226CT14.1.1.2]

Lieferant: United States Biological
USBI030918EA 495 EUR
USBI030918
Anti-DPF2 Mouse Monoclonal Antibody [clone: 226CT14.1.1.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-32000

Type: Primary
Antigen: DPF2 (D4, zinc and double PHD fingers family 2)
Clonality: Monoclonal
Clone: 226CT14.1.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Hamster, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR