Anti-DRD2 Mouse Monoclonal Antibody [clone: 629CT62.2.2]

Lieferant: United States Biological
USBI030922EA 495 EUR
USBI030922
Anti-DRD2 Mouse Monoclonal Antibody [clone: 629CT62.2.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-800

Type: Primary
Antigen: DRD2 (dopamine receptor D2)
Clonality: Monoclonal
Clone: 629CT62.2.2
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR