Anti-DRD4 Mouse Monoclonal Antibody (Biotin) [clone: 625CT1.5.4]

Lieferant: United States Biological
USBI030925-BIOTINEA 659 EUR
USBI030925-BIOTIN
Anti-DRD4 Mouse Monoclonal Antibody (Biotin) [clone: 625CT1.5.4]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: DRD4 (dopamine receptor D4)
Clonality: Monoclonal
Clone: 625CT1.5.4
Conjugation: Biotin
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR