Anti-EIF4E2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 225CT3.1.3]

Lieferant: United States Biological
USBI030938-HRPEA 619 EUR
USBI030938-HRP
Anti-EIF4E2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 225CT3.1.3]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: Monoclonal
Clone: 225CT3.1.3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR