Anti-EIF4EBP1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 319CT22.8.2]

Lieferant: United States Biological
USBI030940-FITCEA 852 EUR
USBI030940-FITC
Anti-EIF4EBP1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 319CT22.8.2]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: Monoclonal
Clone: 319CT22.8.2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR