Anti-ESR1 Mouse Monoclonal Antibody [clone: 415CT16.3.2.1]
Lieferant: United States Biological
USBI030950EA 495 EUR
USBI030950
Anti-ESR1 Mouse Monoclonal Antibody [clone: 415CT16.3.2.1]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-3200
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: Monoclonal
Clone: 415CT16.3.2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human