Anti-GAB1 Mouse Monoclonal Antibody (Biotin) [clone: 1AT1979]

Lieferant: United States Biological
USBI030976-BIOTINEA 720 EUR
USBI030976-BIOTIN
Anti-GAB1 Mouse Monoclonal Antibody (Biotin) [clone: 1AT1979]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-1000

Type: Primary
Antigen: GAB1 (GRB2-associated binding protein 1)
Clonality: Monoclonal
Clone: 1AT1979
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR