Anti-MLST8 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 211CT7.1.1]

Lieferant: United States Biological
USBI030985-FITCEA 616 EUR
USBI030985-FITC
Anti-MLST8 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 211CT7.1.1]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:1560

Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: Monoclonal
Clone: 211CT7.1.1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR