Anti-GSN Mouse Monoclonal Antibody [clone: 328CT2.2.1]

Lieferant: United States Biological
USBI031003EA 495 EUR
USBI031003
Anti-GSN Mouse Monoclonal Antibody [clone: 328CT2.2.1]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: GSN (gelsolin)
Clonality: Monoclonal
Clone: 328CT2.2.1
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR