Anti-Glutathione S Transferase Mouse Monoclonal Antibody (Biotin) [clone: 21CT54.13.1 / 21CT54.13.7]

Lieferant: United States Biological
USBI031004-BIOTINEA 659 EUR
USBI031004-BIOTIN
Anti-Glutathione S Transferase Mouse Monoclonal Antibody (Biotin) [clone: 21CT54.13.1 / 21CT54.13.7]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Tag
Antigen: Glutathione S Transferase
Clonality: Monoclonal
Clone: 21CT54.13.1 / 21CT54.13.7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR