Anti-GSTA1 Mouse Monoclonal Antibody [clone: 286CT8.1.5]

Lieferant: United States Biological
USBI031006EA 495 EUR
USBI031006
Anti-GSTA1 Mouse Monoclonal Antibody [clone: 286CT8.1.5]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-32000

Type: Tag
Antigen: GSTA1 (glutathione S-transferase alpha 1)
Clonality: Monoclonal
Clone: 286CT8.1.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR