Anti-HNF1A Mouse Monoclonal Antibody [clone: 622CT11.9.6]

Lieferant: United States Biological
USBI031029EA 495 EUR
USBI031029
Anti-HNF1A Mouse Monoclonal Antibody [clone: 622CT11.9.6]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: HNF1A (HNF1 homeobox A)
Clonality: Monoclonal
Clone: 622CT11.9.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR