Anti-IGHD Mouse Monoclonal Antibody [clone: 607CT8.4.4]

Lieferant: United States Biological
USBI031042EA 587 EUR
USBI031042
Anti-IGHD Mouse Monoclonal Antibody [clone: 607CT8.4.4]
Antikörper
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: IGHD (immunoglobulin heavy constant delta)
Clonality: Monoclonal
Clone: 607CT8.4.4
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR