Anti-IKBKB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 62AT216]

Lieferant: United States Biological
USBI031045-PEEA 619 EUR
USBI031045-PE
Anti-IKBKB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 62AT216]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive

Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: Monoclonal
Clone: 62AT216
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR