Anti-IKBKG Mouse Monoclonal Antibody [clone: 55AT986.5.78]

Lieferant: United States Biological
USBI031047EA 495 EUR
USBI031047
Anti-IKBKG Mouse Monoclonal Antibody [clone: 55AT986.5.78]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: Monoclonal
Clone: 55AT986.5.78
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR