Anti-NADK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
USBI033002-PEEA 852 EUR
USBI033002-PE
Anti-NADK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NADK2 (NAD kinase 2, mitochondrial)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR